Learn More
Medchemexpress LLC Flavoxate hydrochloride | 3717-88-2 | 99.7% | 250 MG
SDP
Catalog No. 5000405734
Encompass_Preferred
Flavoxate hydrochloride is the analytical standard of Flavoxate (hydrochloride), intended for research and analytical applications. It is a potent and competitive phosphodiesterase (PDE) inhibitor, an antispasmodic agent, and a muscarinic mAChR antagonist. It exhibits moderate calcium antagonistic activity and a local anesthetic effect. This product can be used for the research of overactive bladder (OAB) and lower urinary tract infections.
  • Analytical standard for research and analytical applications.
  • Potent and competitive phosphodiesterase (PDE) inhibitor.
  • Antispasmodic agent.
  • Muscarinic mAChR antagonist.
  • Shows moderate calcium antagonistic activity.
  • Possesses local anesthetic effect.
  • Applicable for research on overactive bladder (OAB) and lower urinary tract infections.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-05734 Supplier Medchemexpress LLC Supplier No. 250MGHYB0549AR
Flavoxate hydrochloride is the analytical standard of Flavoxate (hydrochloride), intended for research and analytical applications. It is a potent and competitive phosphodiesterase (PDE) inhibitor, an antispasmodic agent, and a muscarinic mAChR antagonist. It exhibits moderate calcium antagonistic activity and a local anesthetic effect. This product can be used for the research of overactive bladder (OAB) and lower urinary tract infections.
  • Analytical standard for research and analytical applications.
  • Potent and competitive phosphodiesterase (PDE) inhibitor.
  • Antispasmodic agent.
  • Muscarinic mAChR antagonist.
  • Shows moderate calcium antagonistic activity.
  • Possesses local anesthetic effect.
  • Applicable for research on overactive bladder (OAB) and lower urinary tract infections.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.