Learn More
Medchemexpress LLC Flavoxate hydrochloride | 3717-88-2 | 99.7% | 427.92 | C24H26ClNO4 | 10 MG
SDP
Catalog No. 5000298914
Encompass_Preferred
Flavoxate hydrochloride (standard) is an analytical reference standard supplied for research and analytical applications. It is used as a reference in studies of phosphodiesterase activity and lower urinary tract disorders, and displays antispasmodic, muscarinic antagonist, calcium antagonistic, and local anesthetic properties.
  • Analytical standard for research and method development.
  • High purity suitable for reference and analytical use.
  • Available in small, defined package sizes for assay calibration.
  • Defined chemical identity with molecular weight and formula for traceability.
  • Applicable to studies of overactive bladder and lower urinary tract conditions.
Catalog No. 50-002-98914 Supplier Medchemexpress LLC Supplier No. HYB0549AR10MG
Flavoxate hydrochloride (standard) is an analytical reference standard supplied for research and analytical applications. It is used as a reference in studies of phosphodiesterase activity and lower urinary tract disorders, and displays antispasmodic, muscarinic antagonist, calcium antagonistic, and local anesthetic properties.
  • Analytical standard for research and method development.
  • High purity suitable for reference and analytical use.
  • Available in small, defined package sizes for assay calibration.
  • Defined chemical identity with molecular weight and formula for traceability.
  • Applicable to studies of overactive bladder and lower urinary tract conditions.