Learn More
Medchemexpress LLC Flavoxate hydrochloride | 3717-88-2 | 99.7% | 500 MG
SDP
Catalog No. 5000407886
Encompass_Preferred
Flavoxate hydrochloride is an orally active L-type Ca2+ channel inhibitor and antispasmodic. It inhibits cyclic adenosine monophosphate production by blocking voltage-dependent inward Ba2+ currents, regulating the brainstem micturition center, and stimulating G protein-coupled receptors.
  • Relaxes bladder smooth muscle
  • Inhibits isovolumetric rhythmic contractions
  • Increases bladder capacity
  • Alleviates symptoms of urgent urination frequency and pollakiuria
  • Suitable for research on overactive bladder and related voiding dysfunctions
Catalog No. 50-004-07886 Supplier Medchemexpress LLC Supplier No. HYB0549A500MG
Flavoxate hydrochloride is an orally active L-type Ca2+ channel inhibitor and antispasmodic. It inhibits cyclic adenosine monophosphate production by blocking voltage-dependent inward Ba2+ currents, regulating the brainstem micturition center, and stimulating G protein-coupled receptors.
  • Relaxes bladder smooth muscle
  • Inhibits isovolumetric rhythmic contractions
  • Increases bladder capacity
  • Alleviates symptoms of urgent urination frequency and pollakiuria
  • Suitable for research on overactive bladder and related voiding dysfunctions