Learn More
Medchemexpress LLC Flavoxate hydrochloride | 3717-88-2 | MFCD00072099 | 99.7% | 50 MG
SDP
Catalog No. 5000409432
Encompass_Preferred
Flavoxate hydrochloride is an analytical standard used in research and analytical applications. It acts as a potent and competitive phosphodiesterase (PDE) inhibitor, an antispasmodic agent, and a muscarinic mAChR antagonist. It also exhibits moderate calcium antagonistic activity and a local anesthetic effect.
  • Used for research of overactive bladder (OAB) and lower urinary tract infections.
  • Reference standard for assays.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
Catalog No. 50-004-09432 Supplier Medchemexpress LLC Supplier No. HYB0549AR50MG
Flavoxate hydrochloride is an analytical standard used in research and analytical applications. It acts as a potent and competitive phosphodiesterase (PDE) inhibitor, an antispasmodic agent, and a muscarinic mAChR antagonist. It also exhibits moderate calcium antagonistic activity and a local anesthetic effect.
  • Used for research of overactive bladder (OAB) and lower urinary tract infections.
  • Reference standard for assays.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.