Learn More
Cayman Chemical FlavoxatehydrochlorIde 10mg
SDP
Catalog No. 502983529
Encompass_Preferred
An inhibitor of L-type calcium channels and an antimuscarinic agent; induces relaxation of potassium-precontracted isolated human urinary bladder detrusor muscle (IC50 = 2 µM) and reduces carbachol- or calcium-induced contractions in isolated rat detrusor strips; inhibits reflex micturition in decerebrated cats when microinjected into the oral pontine reticular nucleus at 0.68 UG/animal
Catalog No. 50-298-3529 Supplier Cayman Chemical Supplier No. 1909610 MG
An inhibitor of L-type calcium channels and an antimuscarinic agent; induces relaxation of potassium-precontracted isolated human urinary bladder detrusor muscle (IC50 = 2 µM) and reduces carbachol- or calcium-induced contractions in isolated rat detrusor strips; inhibits reflex micturition in decerebrated cats when microinjected into the oral pontine reticular nucleus at 0.68 UG/animal