Learn More
Cayman Chemical Flavoxate (hydrochloride), Purity: ≥98%, CAS: 3717-88-2, 50 mg
SDP
Catalog No. 502983764
Encompass_Preferred
An inhibitor of L-type calcium channels and an antimuscarinic agent; induces relaxation of potassium-precontracted isolated human urinary bladder detrusor muscle (IC50 = 2 µM) and reduces carbachol- or calcium-induced contractions in isolated rat detrusor strips; inhibits reflex micturition in decerebrated cats when microinjected into the oral pontine reticular nucleus at 0.68 µg/animal
Catalog No. 50-298-3764 Supplier Cayman Chemical Supplier No. 1909650 MG
An inhibitor of L-type calcium channels and an antimuscarinic agent; induces relaxation of potassium-precontracted isolated human urinary bladder detrusor muscle (IC50 = 2 µM) and reduces carbachol- or calcium-induced contractions in isolated rat detrusor strips; inhibits reflex micturition in decerebrated cats when microinjected into the oral pontine reticular nucleus at 0.68 µg/animal