Learn More
Aviva Systems Biology Corp FLCN Antibody - C-terminal region (ARP61521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLCN. It was validated on Western Blot. . Alternate names: BHD, DKFZp547A118, FLCL, FLJ45004, FLJ99377, MGC17998, MGC23445ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-401