Learn More
ABclonal Technology FLCN Rabbit pAb
SDP
Supplier:  ABclonal Technology A145211000UL
Encompass_Preferred
This gene is located within the Smith-Magenis syndrome region on chromosome 17. Mutations in this gene are associated with Birt-Hogg-Dube syndrome, which is characterized by fibrofolliculomas, renal tumors, lung cysts, and pneumothorax. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Catalog No. 50-001-08235