Learn More
Research Products International Corp Flea Micro Magnetic Stirring Bar, 10 x 3 mm
SDP
Catalog No. 501978697
Encompass
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers. Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.
Catalog No. 50-197-8697 Supplier Research Products International Corp Supplier No. F371190010
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers. Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.