Learn More
Cole-Parmer DWK Life Sciences (Kimble) 14035-1000 BEAKERplus Combination Beaker and Flask, 1000 mL, 2/pk
Catalog No. NC1911928 Shop All Cole-Parmer Products
Encompass_Preferred
Kimax BeakerPlus™ Combination Beaker and Flask is ideal for mixing and for use with funnels or filters. The unique design features a wide fluted rim with pour spout and narrow recessed neck to reduce splashing during agitation. Manufactured with borosilicate glass, these beaker/flasks offer excellent chemical and heat resistance.
Catalog No. NC1911928 Supplier Cole-Parmer Supplier No. 3451706
Kimax BeakerPlus™ Combination Beaker and Flask is ideal for mixing and for use with funnels or filters. The unique design features a wide fluted rim with pour spout and narrow recessed neck to reduce splashing during agitation. Manufactured with borosilicate glass, these beaker/flasks offer excellent chemical and heat resistance.