Learn More
Medchemexpress LLC Flecainide | 54143-55-4 | 100.0% | 414.34 | 100 MG
SDP
Catalog No. 5000366462
Encompass_Preferred
Flecainide is an orally active antiarrhythmic agent that blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It is used in research for diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Orally active antiarrhythmic agent.
  • Blocks sodium channels.
  • Inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2).
  • Used in research for diseases such as catecholaminergic ventricular tachycardia (CPVT).
  • In vitro studies show it significantly reduces irregular Ca2+ release in single cardiomyocytes.
  • Under investigation for premature ventricular beats, premature ventricular complexes, and various ventricular arrhythmias.
  • Clinical trials are exploring its use for atrial fibrillation and supraventricular tachycardia.
  • Has been studied for pain management.
Catalog No. 50-003-66462 Supplier Medchemexpress LLC Supplier No. HYW010950100MG
Flecainide is an orally active antiarrhythmic agent that blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It is used in research for diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Orally active antiarrhythmic agent.
  • Blocks sodium channels.
  • Inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2).
  • Used in research for diseases such as catecholaminergic ventricular tachycardia (CPVT).
  • In vitro studies show it significantly reduces irregular Ca2+ release in single cardiomyocytes.
  • Under investigation for premature ventricular beats, premature ventricular complexes, and various ventricular arrhythmias.
  • Clinical trials are exploring its use for atrial fibrillation and supraventricular tachycardia.
  • Has been studied for pain management.