Learn More
Medchemexpress LLC Flecainide | 54143-55-4 | 414.34 | 10 MG
Supplier: Medchemexpress LLC HYW01095010MG
Flecainide (Standard) is the analytical standard of Flecainide, intended for research and analytical applications. It functions as an orally active antiarrhythmic agent by blocking sodium channels and inhibiting calcium ion release mediated by the cardiac ryanodine receptor (RyR2). This product is suitable for research into diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
- Used in qualitative, quantitative, and methodological research experiments
- Suitable for HPLC, GC, and MS applications
- Blocks sodium channels
- Inhibits calcium ion release mediated by cardiac ryanodine receptor (RyR2)
- Applicable for cardiovascular disease and blood or cardio-cerebrovascular disease research