Learn More
Medchemexpress LLC Flecainide | 54143-55-4 | 414.34 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01095050MG
Encompass_Preferred
Flecainide (Standard) is the analytical standard of Flecainide, intended for research and analytical applications. Flecainide is an orally active antiarrhythmic agent that blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It can be used in the research of diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Analytical standard for research and analytical applications
  • Orally active antiarrhythmic agent
  • Blocks sodium channels
  • Inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2)
  • Useful in research of diseases like catecholaminergic polymorphic ventricular tachycardia (CPVT)
  • Reference standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-67028