Learn More
Medchemexpress LLC Flecainide | 54143-55-4 | >99.9% | 414.34 | 5 MG
SDP
Catalog No. 5000452270
Encompass_Preferred
Flecainide is an orally active antiarrhythmic agent. It blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It is used in research for diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Blocks sodium channels.
  • Inhibits calcium ion release mediated by cardiac ryanodine receptor (RyR2).
  • Used in research for catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • 99.98% purity as a solid.
Catalog No. 50-004-52270 Supplier Medchemexpress LLC Supplier No. HYW0109505MG
Flecainide is an orally active antiarrhythmic agent. It blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It is used in research for diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Blocks sodium channels.
  • Inhibits calcium ion release mediated by cardiac ryanodine receptor (RyR2).
  • Used in research for catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • 99.98% purity as a solid.