Learn More
TARGETMOL CHEMICALS INC FLECAINIDE ACETATE 50MG
SDP
Catalog No. 502579235
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Flecainide acetate (R-818) is a class Ic antiarrhythmic agent used to prevent and treat tachyarrhythmias (abnormal fast rhythms of the heart). purity: 99%
Catalog No. 50-257-9235 Supplier TARGETMOL CHEMICALS INC Supplier No. T721950MG
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Flecainide acetate (R-818) is a class Ic antiarrhythmic agent used to prevent and treat tachyarrhythmias (abnormal fast rhythms of the heart). purity: 99%