Learn More
Medchemexpress LLC Flecainide (acetate) | 54143-56-5 | 99.4% | 474.39 | 50 MG
SDP
Catalog No. 5000386243
Encompass_Preferred
Flecainide acetate (R-818) is a class 1C antiarrhythmic agent primarily used for the management of supraventricular arrhythmia. It functions by blocking the Nav1.5 sodium channel in the heart, leading to the prolongation of the cardiac action potential.
  • Class 1C antiarrhythmic agent
  • Blocks the Nav1.5 sodium channel
  • Causes prolongation of the cardiac action potential
  • For research use only
Catalog No. 50-003-86243 Supplier Medchemexpress LLC Supplier No. HY1742950MG
Flecainide acetate (R-818) is a class 1C antiarrhythmic agent primarily used for the management of supraventricular arrhythmia. It functions by blocking the Nav1.5 sodium channel in the heart, leading to the prolongation of the cardiac action potential.
  • Class 1C antiarrhythmic agent
  • Blocks the Nav1.5 sodium channel
  • Causes prolongation of the cardiac action potential
  • For research use only