Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flecainide acetate | 54143-56-5 | MFCD00214290 | 99.4% | 474.39 g/mol | C19H24F6N2O5 | 10 MG
SDP

Catalog No. 5000299102
Encompass_Preferred

Flecainide acetate is a class 1C antiarrhythmic research reagent that inhibits cardiac Nav1.5 sodium channels. Supplied as a white to off-white solid for laboratory use; intended for research use only and not for human or clinical use.

  • Class 1C antiarrhythmic agent that blocks Nav1.5 sodium channels.
  • Supplied as a white to off-white solid suitable for analytical and biological research.
  • High purity of 99.4%.
  • Molecular formula C19H24F6N2O5; molecular weight 474.39 g/mol.
  • Available in small research pack sizes, including 10 mg.
  • Store solid at 4°C; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-99102 Supplier Medchemexpress LLC Supplier No. HY1742910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flecainide acetate is a class 1C antiarrhythmic research reagent that inhibits cardiac Nav1.5 sodium channels. Supplied as a white to off-white solid for laboratory use; intended for research use only and not for human or clinical use.

  • Class 1C antiarrhythmic agent that blocks Nav1.5 sodium channels.
  • Supplied as a white to off-white solid suitable for analytical and biological research.
  • High purity of 99.4%.
  • Molecular formula C19H24F6N2O5; molecular weight 474.39 g/mol.
  • Available in small research pack sizes, including 10 mg.
  • Store solid at 4°C; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.