Learn More
Medchemexpress LLC Flecainide acetate (R-818) | 54143-56-5 | 99.4% | 474.39 | 1 ML
SDP
Catalog No. 5000386611
Encompass_Preferred
Flecainide acetate (R-818) is a class 1C antiarrhythmic agent primarily used for managing supraventricular arrhythmia. It functions by blocking the Nav1.5 sodium channel in the heart, which results in the prolongation of the cardiac action potential. This product is for research use only.
  • It is a class 1C antiarrhythmic agent.
  • It is especially used for the management of supraventricular arrhythmia.
  • It works by blocking the Nav1.5 sodium channel in the heart.
  • It causes prolongation of the cardiac action potential.
Catalog No. 50-003-86611 Supplier Medchemexpress LLC Supplier No. HY1742910MM/1ML
Flecainide acetate (R-818) is a class 1C antiarrhythmic agent primarily used for managing supraventricular arrhythmia. It functions by blocking the Nav1.5 sodium channel in the heart, which results in the prolongation of the cardiac action potential. This product is for research use only.
  • It is a class 1C antiarrhythmic agent.
  • It is especially used for the management of supraventricular arrhythmia.
  • It works by blocking the Nav1.5 sodium channel in the heart.
  • It causes prolongation of the cardiac action potential.