Learn More
Medchemexpress LLC Flecainide hydrochloride | 57415-44-8 | 99.93% | 450.80 | 1 ML
SDP
Catalog No. 5000366279
Encompass_Preferred
Flecainide hydrochloride is an orally active antiarrhythmic agent that blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It can be used in the research of diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Significantly reduces irregular Ca2+ release in single cardiomyocytes derived from ATS-iPSCs.
  • Completely inhibits catecholamine-induced ventricular arrhythmias in Casq2 knockout mice.
Catalog No. 50-003-66279 Supplier Medchemexpress LLC Supplier No. HYW010950A10MM/1ML
Flecainide hydrochloride is an orally active antiarrhythmic agent that blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It can be used in the research of diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Significantly reduces irregular Ca2+ release in single cardiomyocytes derived from ATS-iPSCs.
  • Completely inhibits catecholamine-induced ventricular arrhythmias in Casq2 knockout mice.