Learn More
Medchemexpress LLC Flecainide hydrochloride | 57415-44-8 | 99.93% | 450.80 | 100 MG
SDP
Catalog No. 5000451969
Encompass_Preferred
Flecainide hydrochloride is an orally active antiarrhythmic agent. It blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It can be used in the research of diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • In vitro: flecainide (hydrochloride) (500 nM) significantly reduces irregular Ca2+ release in single cardiomyocytes derived from ATS-iPSCs.
  • In vivo: flecainide (hydrochloride) (20 mg/kg; intraperitoneal injection; single dose) completely inhibits catecholamine-induced ventricular arrhythmias in Casq2 knockout mice.
Catalog No. 50-004-51969 Supplier Medchemexpress LLC Supplier No. HYW010950A100MG
Flecainide hydrochloride is an orally active antiarrhythmic agent. It blocks sodium channels and inhibits calcium ion release mediated by the cardiac ryanodine receptor (RyR2). It can be used in the research of diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • In vitro: flecainide (hydrochloride) (500 nM) significantly reduces irregular Ca2+ release in single cardiomyocytes derived from ATS-iPSCs.
  • In vivo: flecainide (hydrochloride) (20 mg/kg; intraperitoneal injection; single dose) completely inhibits catecholamine-induced ventricular arrhythmias in Casq2 knockout mice.