Learn More
Cayman Chemical FlecaInIdeacetate 10mg
SDP
Supplier:  Cayman Chemical 2038810 MG
Encompass_Preferred
An inhibitor of cardiac late INa channels (IC50 = 3.4 µM) and IKr channels (IC50 = 1.5 µM); used clinically to suppress arrhythmias and sodium-dependent calcium overload associated with myocardial ischemia and heart failure
Catalog No. 50-298-4482