Learn More
Cayman Chemical Flecainide (acetate), Purity: ≥98%, CAS: 54143-56-5, 50 mg
SDP
Catalog No. 502984604
Encompass_Preferred
An inhibitor of cardiac late INa channels (IC50 = 3.4 µM) and IKr channels (IC50 = 1.5 µM); used clinically to suppress arrhythmias and sodium-dependent calcium overload associated with myocardial ischemia and heart failure
Catalog No. 50-298-4604 Supplier Cayman Chemical Supplier No. 2038850 MG
An inhibitor of cardiac late INa channels (IC50 = 3.4 µM) and IKr channels (IC50 = 1.5 µM); used clinically to suppress arrhythmias and sodium-dependent calcium overload associated with myocardial ischemia and heart failure