Learn More
National Safety Apparel™ Fleece FR Hard Hat Liner
SDP
Catalog No. 17001743 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-743 Color: Navy
1 options
Catalog No. 17-001-743 Supplier National Safety Apparel™ Supplier No. H74FL10
Features FR hook and loop closure and hook and loop tabs to attach to your hard hat. Made from Dupont Nomex fleece, this liner was made to deliver superior warmth in cold climates.
  • Arc Rating: 24 cal/cm²
  • CAT 2
  • Fabric/Material: 8.5 oz. DuPont™ Nomex™ IIIA Fleece
  • Made from Dupont™ Nomex™ fleece
  • Superior warmth in cold climates
  • Warmth without the weight FR hook and loop closure
  • Hook and loop tabs to attach to hard hat
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™, Kevlar™, Anti-Stat
Color Navy
Closure Type FR Hook and Loop
Seam Type Stitched
Arc Flash Rating 24 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, ASTM F1506, CSA Z462, OSHA 1910.269, NFPA 2112