Learn More
MSC Viking Firewall Bib Overalls, 88% Cotton, 12% Nylon, 12.4 cal/cm² ARC, 1 ea
Catalog No. 5000688768
Encompass
These flame-resistant bib overalls are engineered for dual-hazard protection against electrical arc and hydrocarbon flash fire. Constructed from 9 oz. Westex® by Milliken® 88% cotton and 12% nylon fabric, they provide an arc thermal performance value of 12.4 cal/cm² (CAT 2). The overalls are designed for durability in industrial environments.
  • Meets NFPA 2112, CSA Z462, ASTM F1506, and NFPA 70E safety standards
  • Features 3M Scotchlite™ flame-resistant reflective trim (2 in. yellow/silver/yellow)
  • High visibility orange complies with CSA Z96-22 Class 2, Level FR
  • Includes two-way breakaway Nomex® backed YKK® front zipper with exterior storm flap
  • Equipped with adjustable shoulder suspenders, multiple storage pockets, a tool pocket, and a hammer loop
  • Double-stitched seams with bar-tacked stress points
Catalog No. 50-006-88768 Supplier MSC Supplier No. 93254928
These flame-resistant bib overalls are engineered for dual-hazard protection against electrical arc and hydrocarbon flash fire. Constructed from 9 oz. Westex® by Milliken® 88% cotton and 12% nylon fabric, they provide an arc thermal performance value of 12.4 cal/cm² (CAT 2). The overalls are designed for durability in industrial environments.
  • Meets NFPA 2112, CSA Z462, ASTM F1506, and NFPA 70E safety standards
  • Features 3M Scotchlite™ flame-resistant reflective trim (2 in. yellow/silver/yellow)
  • High visibility orange complies with CSA Z96-22 Class 2, Level FR
  • Includes two-way breakaway Nomex® backed YKK® front zipper with exterior storm flap
  • Equipped with adjustable shoulder suspenders, multiple storage pockets, a tool pocket, and a hammer loop
  • Double-stitched seams with bar-tacked stress points