Learn More
Analytical Sales & Services Fleet™ Armor™ C18 Column. 5µm, 10mm x 1.0mm ID
Catalog No. 5000806827
Encompass
Fleet™ Armor™ C18 Column. 5µm, 10mm x 1.0mm ID, 6-32 threads for capillary fittings. Monofunctional with unique polar and bulky end-capping.
Catalog No. 50-008-06827 Supplier Analytical Sales & Services Supplier No. AT1811
Fleet™ Armor™ C18 Column. 5µm, 10mm x 1.0mm ID, 6-32 threads for capillary fittings. Monofunctional with unique polar and bulky end-capping.