Learn More
Analytical Sales & Services Fleet™ Armor™ C18 Column. 5µm, 2.5mm x 0.5mm ID
Catalog No. 5000804893
Encompass
Fleet™ Armor™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. Monofunctional with unique polar and bulky end-capping.
Catalog No. 50-008-04893 Supplier Analytical Sales & Services Supplier No. AT1825
Fleet™ Armor™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. Monofunctional with unique polar and bulky end-capping.