Learn More
Analytical Sales & Services Fleet™ Armor™ C18 Column. 5µm, 5mm x 1.0mm ID
Catalog No. 5000806828
Encompass
Fleet™ Armor™ C18 Column. 5µm, 5mm x 1.0mm ID, 6-32 Threads for Capillary Tubing. Monofunctional with unique polar and bulky end-capping.
Catalog No. 50-008-06828 Supplier Analytical Sales & Services Supplier No. AT1851
Fleet™ Armor™ C18 Column. 5µm, 5mm x 1.0mm ID, 6-32 Threads for Capillary Tubing. Monofunctional with unique polar and bulky end-capping.