Learn More
Analytical Sales & Services Fleet™ Echelon™ C18 Column. 5µm, 10mm x 1.0mm ID
Catalog No. 5000806416
Encompass
Fleet™ Echelon™ C18 Column. 5µm, 10mm x 1.0mm ID, 6-32 threads for capillary fittings. Designed for high resolution and selectivity.
Catalog No. 50-008-06416 Supplier Analytical Sales & Services Supplier No. AE1811
Fleet™ Echelon™ C18 Column. 5µm, 10mm x 1.0mm ID, 6-32 threads for capillary fittings. Designed for high resolution and selectivity.