Learn More
Analytical Sales & Services Fleet™ Echelon™ C18 Column. 5µm, 2.5mm x 0.5mm ID
Catalog No. 5000805971
Encompass
Fleet™ Echelon™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. Designed for high resolution and selectivity.
Catalog No. 50-008-05971 Supplier Analytical Sales & Services Supplier No. AE1825
Fleet™ Echelon™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. Designed for high resolution and selectivity.