Learn More
Analytical Sales & Services Fleet™ Highload™ C18 Column. 5µm, 10mm x 1.0mm ID
Catalog No. 5000805595
Encompass
Fleet™ Highload™ C18 Column. 5µm, 10mm x 1.0mm ID, 6-32 threads for capillary fittings. For applications requiring maximum loading and selectivity.
Catalog No. 50-008-05595 Supplier Analytical Sales & Services Supplier No. HT1811
Fleet™ Highload™ C18 Column. 5µm, 10mm x 1.0mm ID, 6-32 threads for capillary fittings. For applications requiring maximum loading and selectivity.