Learn More
Analytical Sales & Services Fleet™ Highload™ C18 Column. 5µm, 2.5mm x 0.5mm ID
Catalog No. 5000806616
Encompass
Fleet™ Highload™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. For applications requiring maximum loading and selectivity.
Catalog No. 50-008-06616 Supplier Analytical Sales & Services Supplier No. HT1825
Fleet™ Highload™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. For applications requiring maximum loading and selectivity.