Learn More
Analytical Sales & Services Fleet™ Lancer™ C18 Column. 5µm, 2.5mm x 0.5mm ID
Catalog No. 5000805196
Encompass
Fleet™ Lancer™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. Extremely well end-capped monofunctional phase.
Catalog No. 50-008-05196 Supplier Analytical Sales & Services Supplier No. LT1825
Fleet™ Lancer™ C18 Column. 5µm, 2.5mm x 0.5mm ID, 6-32 threads for capillary fittings. Extremely well end-capped monofunctional phase.