Learn More
Analytical Sales & Services Fleet™ Lancer™ C18 Column. 5µm, 5mm x 1.0mm ID
Catalog No. 5000805193
Encompass
Fleet™ Lancer™ C18 Column. 5µm, 5mm x 1.0mm ID, 6-32 threads for capillary fittings. Extremely well end-capped monofunctional phase.
Catalog No. 50-008-05193 Supplier Analytical Sales & Services Supplier No. LT1851
Fleet™ Lancer™ C18 Column. 5µm, 5mm x 1.0mm ID, 6-32 threads for capillary fittings. Extremely well end-capped monofunctional phase.