Learn More
Medchemexpress LLC Fleroxacin-d3 hydrochloride | 79660-72-3 | 98.8% | 1 MG
SDP
Catalog No. 5000408584
Encompass_Preferred
Fleroxacin-d3 (hydrochloride) is the deuterium labeled Fleroxacin (hydrochloride). It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for various analyses.
  • Functions as an internal standard for quantitative analysis.
  • Useful for NMR, GC-MS, or LC-MS applications.
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08584 Supplier Medchemexpress LLC Supplier No. HYB0414AS1MG
Fleroxacin-d3 (hydrochloride) is the deuterium labeled Fleroxacin (hydrochloride). It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for various analyses.
  • Functions as an internal standard for quantitative analysis.
  • Useful for NMR, GC-MS, or LC-MS applications.
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs.