Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fleroxacin-d3 hydrochloride | 79660-72-3 | 98.8% | 5 MG
SDP

Catalog No. 5000409242
Encompass_Preferred

Fleroxacin-d3 hydrochloride is the deuterium-labeled form of Fleroxacin hydrochloride. This compound is used as a tracer and as an internal standard for quantitative analysis in various analytical methods. Deuteration, the incorporation of stable heavy isotopes, offers potential benefits by affecting the pharmacokinetic and metabolic profiles of drugs, making it valuable in drug development.

  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight: 408.82
  • Formula: C17H16D3ClF3N3O3
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-09242 Supplier Medchemexpress LLC Supplier No. HYB0414AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fleroxacin-d3 hydrochloride is the deuterium-labeled form of Fleroxacin hydrochloride. This compound is used as a tracer and as an internal standard for quantitative analysis in various analytical methods. Deuteration, the incorporation of stable heavy isotopes, offers potential benefits by affecting the pharmacokinetic and metabolic profiles of drugs, making it valuable in drug development.

  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight: 408.82
  • Formula: C17H16D3ClF3N3O3
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.