Learn More
Medchemexpress LLC Flesinoxan | 98206-10-1 | 99.9% | 10 MG
SDP
Catalog No. 5000232477
Encompass_Preferred
Flesinoxan is a potent, selective 5-hydroxytryptamine 1A (5-HT1A) receptor agonist used for pharmacological research. Supplied as a high-purity solid, it is suitable for in vitro and in vivo studies when handled and stored per recommended conditions.
  • Potent, high-affinity 5-HT1A receptor agonist.
  • High purity suitable for research applications.
  • Soluble in DMSO; in vivo formulations documented.
  • Stable under recommended storage conditions.
  • Available in small research-scale pack sizes.
Catalog No. 50-002-32477 Supplier Medchemexpress LLC Supplier No. HY12165310MG
Flesinoxan is a potent, selective 5-hydroxytryptamine 1A (5-HT1A) receptor agonist used for pharmacological research. Supplied as a high-purity solid, it is suitable for in vitro and in vivo studies when handled and stored per recommended conditions.
  • Potent, high-affinity 5-HT1A receptor agonist.
  • High purity suitable for research applications.
  • Soluble in DMSO; in vivo formulations documented.
  • Stable under recommended storage conditions.
  • Available in small research-scale pack sizes.