Learn More
Medchemexpress LLC Flesinoxan | 98206-10-1 | 99.9% | 100 MG
SDP
Catalog No. 5000231602
Encompass_Preferred
Flesinoxan is a selective 5-hydroxytryptamine 1A (5-HT1A) receptor agonist provided as a high-purity research standard. It is intended for receptor pharmacology, in vitro assays, and analytical applications; molecular formula C22H26FN3O4, molecular weight 415.46.
  • Selective 5-HT1A receptor agonist for pharmacology studies.
  • High reported purity (99.9%) suitable for analytical applications.
  • Available in research-scale pack sizes for laboratory use.
  • Provided as a DMSO solution option for assay preparation.
  • Suitable as an analytical standard for in vitro assays.
Catalog No. 50-002-31602 Supplier Medchemexpress LLC Supplier No. HY121653100MG
Flesinoxan is a selective 5-hydroxytryptamine 1A (5-HT1A) receptor agonist provided as a high-purity research standard. It is intended for receptor pharmacology, in vitro assays, and analytical applications; molecular formula C22H26FN3O4, molecular weight 415.46.
  • Selective 5-HT1A receptor agonist for pharmacology studies.
  • High reported purity (99.9%) suitable for analytical applications.
  • Available in research-scale pack sizes for laboratory use.
  • Provided as a DMSO solution option for assay preparation.
  • Suitable as an analytical standard for in vitro assays.