Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flesinoxan | 98206-10-1 | 99.9% | 50 MG
SDP

Catalog No. 5000232274
Encompass_Preferred

Flesinoxan is a selective 5-HT1A receptor agonist research compound intended for laboratory research use. It is supplied as an off-white to light yellow solid with demonstrated potency at the 5-HT1A receptor and standard handling and storage recommendations for small-molecule research chemicals.

  • Selective 5-HT1A receptor agonist (EC50 24 nM).
  • High reported purity: 99.9%.
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO (31.25 mg/mL); in vivo formulations reported at ≥2.08 mg/mL.
  • Storage: powder stable at -20°C (3 years) and 4°C (2 years).
  • For research use only; not for human consumption or clinical use.

Catalog No. 50-002-32274 Supplier Medchemexpress LLC Supplier No. HY12165350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flesinoxan is a selective 5-HT1A receptor agonist research compound intended for laboratory research use. It is supplied as an off-white to light yellow solid with demonstrated potency at the 5-HT1A receptor and standard handling and storage recommendations for small-molecule research chemicals.

  • Selective 5-HT1A receptor agonist (EC50 24 nM).
  • High reported purity: 99.9%.
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO (31.25 mg/mL); in vivo formulations reported at ≥2.08 mg/mL.
  • Storage: powder stable at -20°C (3 years) and 4°C (2 years).
  • For research use only; not for human consumption or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.