Learn More
Opentrons Labworks Inc FLEX 200UL CLEAR 1920TIPS PK
Catalog No. NC3346583
Encompass
Opentrons Flex Filter Tips, 200 µL 200 µL filtered pipette tips for the Opentrons Flex. Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable. 20 racks
Catalog No. NC3346583 Supplier Opentrons Labworks Inc Supplier No. 99100105
Opentrons Flex Filter Tips, 200 µL 200 µL filtered pipette tips for the Opentrons Flex. Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable. 20 racks