Learn More
Medicus Health Flex Arm Splash Shield with 24 Inch Black Gooseneck Arm
SDP
Catalog No. NC3904715
Encompass
• Flex Arm Safety Splash Shield with 24" Black Gooseneck Arm and C-clamp mounting base
• Flat Splash Shield mounted on a 24" long black vinyl coated flexible goose arm offering flexibility for optimum protection and precise positioning
• Flat 10”L x 10”W flat shield is mounted diagonally so it adds another 13” to the overall length of the shield.
• Coated arm is easy to clean and keep clean
• Durable, scratch-resistant polycarbonate shield protects you from splashes, aerosol mist, and flying debris
• Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)
• C-clamp mount is padded to protect your desk or bench top from scratches and can be mounted on a surface up to 2" thick
• To create a permanent mount, simply remove the C-clamp and fasten to any solid surface
Catalog No. NC3904715 Supplier Medicus Health Supplier No. 5270M31
• Flex Arm Safety Splash Shield with 24" Black Gooseneck Arm and C-clamp mounting base
• Flat Splash Shield mounted on a 24" long black vinyl coated flexible goose arm offering flexibility for optimum protection and precise positioning
• Flat 10”L x 10”W flat shield is mounted diagonally so it adds another 13” to the overall length of the shield.
• Coated arm is easy to clean and keep clean
• Durable, scratch-resistant polycarbonate shield protects you from splashes, aerosol mist, and flying debris
• Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)
• C-clamp mount is padded to protect your desk or bench top from scratches and can be mounted on a surface up to 2" thick
• To create a permanent mount, simply remove the C-clamp and fasten to any solid surface