Learn More
Thermo Scientific Flex Cap Viper MP35N 1pc
Supplier:  Thermo Scientific 6041.7892
Encompass
A flexible MP35N nickel-cobalt-chromium-molybdenum alloy cap that seals injection ports and connectors on gas chromatography columns and instruments, replacing part 6041.7892 on specified gas chromatography systems.
Catalog No. NC1272682