Learn More
Electron Microscopy Sciences Flex Conductive Pen, Regular Tip
SDP
Catalog No. 50980318
Encompass_Preferred
This micro-tip pen is used for drawing flexible conductive silver traces on Mylar and any flex circuitry. Dries in minutes.
Catalog No. 50-980-318 Supplier Electron Microscopy Sciences Supplier No. 1264501
This micro-tip pen is used for drawing flexible conductive silver traces on Mylar and any flex circuitry. Dries in minutes.