Learn More
Ace Glass, Inc. 3/8" flex coupling for 10mm stir shafts
SDP
Catalog No. 50976099
Encompass
3/8" flex coupling for 10mm stir shafts consists of a flexible neoprene rubber body with a PTFE sleeve inside a corrosion-resistant metal end. Flex coupling provides angular and parallel misalignment of glass or metal stirring shaft, without danger of breakage. Rubber body absorbs shock and provides quiet, vibration-free running with great torsional stiffness. Measures 4 overall.
Catalog No. 50-976-099 Supplier Ace Glass, Inc. Supplier No. 812511
3/8" flex coupling for 10mm stir shafts consists of a flexible neoprene rubber body with a PTFE sleeve inside a corrosion-resistant metal end. Flex coupling provides angular and parallel misalignment of glass or metal stirring shaft, without danger of breakage. Rubber body absorbs shock and provides quiet, vibration-free running with great torsional stiffness. Measures 4 overall.