Learn More
Research Products International Corp Flexible Lead Foil, .030 Inches Thick, 14 Inches Wide, 5 Feet Long
SDP
Catalog No. 50550141
Encompass
Flexible lead foil is ideal for wrapping around tubes, bottles or containers that require shielding from 125I. Clean and easy to use lead foil can be cut with a utility knife or scissors.
Catalog No. 50-550-141 Supplier Research Products International Corp Supplier No. 104030
Flexible lead foil is ideal for wrapping around tubes, bottles or containers that require shielding from 125I. Clean and easy to use lead foil can be cut with a utility knife or scissors.