Learn More
Pyramex™ Flex-Lyte™ Safety Glasses
Catalog No. 19555700 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black, Gray
Black, Lime
Lens Tint:
Clear
Gray
Indoor/Outdoor, Mirror
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-555-700 Black, Lime Indoor/Outdoor, Mirror
19-555-683 Black, Gray Clear
19-555-685 Black, Gray Gray
19-555-687 Black, Gray Indoor/Outdoor, Mirror
19-555-696 Black, Lime Clear
19-555-698 Black, Lime Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-555-700 Supplier Pyramex™ Supplier No. SBL10580D
This item is not returnable. View return policy
Pyramex™ Flex-Lyte™ Safety Glasses with ANSI Z87 safety compliance
  • Soft nose piece for snug fit
  • Semi-frame & slim co-material temple design for lightweight spectacle - 22 gm
  • Offers a clear, panoramic view without obstruction
  • 9 base wraparound lens provides full front side protection
  • Scratch-resistant, polycarbonate lens
  • Lens Technology:
    • Visible Light Transmitted - 96%
    • UVA, UVB Blocked - 100%
    • Blue Light Blocked - 9%
    • IR Blocked - 18%
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87
Description Indoor/Outdoor Mirror Lens
Lens Tint Indoor/Outdoor, Mirror
For Use With (Application) Eye Safety
Frame Color Black, Lime
Temple Style Slim Co-Injected
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400