Learn More
Pyramex™ Flex-Lyte™ Safety Glasses with H2MAX Lens
Catalog No. 19555684 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black, Gray
Black, Lime
Lens Tint:
Clear
Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-555-684 Black, Gray Clear
19-555-686 Black, Gray Gray
19-555-697 Black, Lime Clear
19-555-699 Black, Lime Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-555-684 Supplier Pyramex™ Supplier No. SBG10510DTM
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Pyramex™ Flex-Lyte™ Safety Glasses with ANSI Z87 safety compliance
  • Soft nose piece for snug fit
  • Semi-frame & slim co-material temple design for lightweight spectacle - 22 gm
  • Offers a clear, panoramic view without obstruction
  • 9 base wraparound lens provides full front side protection
  • Scratch-resistant, polycarbonate lens
  • Lens Technology:
    • Visible Light Transmitted - 96%
    • UVA, UVB Blocked - 100%
    • Blue Light Blocked - 9%
    • IR Blocked - 18%
Specifications
Lens Coating Anti-fog
Lens Type H2MAX
Certifications/Compliance ANSI Z87
Description Clear/H2MAX Lens
Lens Tint Clear
For Use With (Application) Eye Safety
Frame Color Black, Gray
Temple Style Slim Co-Injected
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 1