Learn More
Grainger Flex Metal Conduit Steel Trd Sz 3/4in
Catalog No. 502765161
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is sunlight, oil, and chemical resistant. It meets NEC article 350 requirements and is designed for use with flexible metal conduit fittings.
Catalog No. 50-276-5161 Supplier Grainger Supplier No. 16R025
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is sunlight, oil, and chemical resistant. It meets NEC article 350 requirements and is designed for use with flexible metal conduit fittings.