Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1 1/2in
Catalog No. 502768452
Encompass_Preferred
The Grainger 16R058 is a Westward brand 1/4-inch drive socket set. It contains 14 SAE sockets ranging from 1/4 inch to 9/16 inch and 14 metric sockets ranging from 4mm to 14mm. The sockets have a polished chrome finish and are made from chrome vanadium steel for durability. This 108-piece set comes organized in a molded case for convenient storage and portability. The sockets have knurled sides for easy gripping and removal.
Catalog No. 50-276-8452 Supplier Grainger Supplier No. 16R058
The Grainger 16R058 is a Westward brand 1/4-inch drive socket set. It contains 14 SAE sockets ranging from 1/4 inch to 9/16 inch and 14 metric sockets ranging from 4mm to 14mm. The sockets have a polished chrome finish and are made from chrome vanadium steel for durability. This 108-piece set comes organized in a molded case for convenient storage and portability. The sockets have knurled sides for easy gripping and removal.