Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1/2in
Catalog No. 502768519
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a UL listed 360 degree ground and is watertight to protect wires. It comes in 10 foot lengths in trade sizes 3/8 inch to 4 inch. The conduit can be used for exposed runs connecting equipment in damp or wet locations. It is very flexible and easy to install around obstacles.
Catalog No. 50-276-8519 Supplier Grainger Supplier No. 16R010
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a UL listed 360 degree ground and is watertight to protect wires. It comes in 10 foot lengths in trade sizes 3/8 inch to 4 inch. The conduit can be used for exposed runs connecting equipment in damp or wet locations. It is very flexible and easy to install around obstacles.