Learn More
Grainger Flex Metal Conduit Steel Trd Sz 3/4in
Catalog No. 502765163
Encompass_Preferred
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It has an oil-resistant thermoplastic jacket over an interlocked steel construction. This 3/4" trade size conduit can be easily bent and shaped on site for routing over, under and around obstructions. It is UL listed and CSA certified. The working temperature range is -20°F to 105°F. This 25 foot length of flexible metal conduit weighs approximately 6 lbs.
Catalog No. 50-276-5163 Supplier Grainger Supplier No. 16R012
The Flexible Metal Conduit Reduced from Grainger is a flexible metal conduit that provides protection for electrical wiring in exposed locations. It has an oil-resistant thermoplastic jacket over an interlocked steel construction. This 3/4" trade size conduit can be easily bent and shaped on site for routing over, under and around obstructions. It is UL listed and CSA certified. The working temperature range is -20°F to 105°F. This 25 foot length of flexible metal conduit weighs approximately 6 lbs.